Assay Details
| Gene Symbol: GATAD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001266151.1 | NP_001253080.1 | 706 | 100 | ||
| GenBank mRNA | JV047782.1 | - | 706 | 100 | ||
| JV657483.1 | - | 275 | 100 | |||
More Information
Set Membership: |
|

GATAD1
Genomic Map