Assay Details
| Gene Symbol: LURAP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001266426.2 | NP_001253355.1 | 262 | 68 | ||
| GenBank mRNA | JU335054.1 | - | 288 | 68 | ||
| JU476055.1 | - | 251 | 68 | |||
| JV047416.1 | - | 265 | 68 | |||
| JV584832.1 | - | 288 | 68 | |||
| JV654022.1 | - | 290 | 68 | |||
More Information
Set Membership: |
|

LURAP1
Genomic Map