Assay Details
| Gene Symbol: LRRC4B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001266779.1 | NP_001253708.1 | 435 | 73 | ||
| XM_028838522.1 | XP_028694355.1 | 1242 | 73 | |||
| GenBank mRNA | JV633834.1 | - | 402 | 73 | ||
| JV667506.1 | - | 435 | 73 | |||
More Information
Set Membership: |
|

LRRC4B
Genomic Map