Assay Details
| Gene Symbol: GLTP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001260790.1 | NP_001247719.1 | 510 | 72 | ||
| XM_015152734.2 | XP_015008220.1 | 513 | 72 | |||
| GenBank mRNA | JU334826.1 | - | 507 | 72 | ||
| JU334827.1 | - | 507 | 72 | |||
| JU334828.1 | - | 507 | 72 | |||
| JU334829.1 | - | 507 | 72 | |||
| JU334830.1 | - | 507 | 72 | |||
| JU334831.1 | - | 507 | 72 | |||
| JU475858.1 | - | 532 | 72 | |||
| JV043450.1 | - | 510 | 72 | |||
More Information
Set Membership: |
|

GLTP
Genomic Map