Assay Details
| Gene Symbol: OPA3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001258146.2 | NP_001245075.1 | 1016 | 96 | ||
| GenBank mRNA | JU477300.1 | - | 1016 | 96 | ||
| JV636387.1 | - | 1068 | 96 | |||
More Information
Set Membership: |
|

OPA3
Genomic Map