Assay Details
| Gene Symbol: RRP15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015116097.2 | XP_014971583.1 | 456 | 83 | ||
| XM_015116105.2 | XP_014971591.1 | 458 | 83 | |||
| GenBank mRNA | JV652897.1 | - | 417 | 83 | ||
| JV716221.1 | - | 418 | 83 | |||
More Information
Set Membership: |
|

RRP15
Genomic Map