Assay Details
| Gene Symbol: ZNF432 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015124738.2 | XP_014980224.1 | 220 | 104 | ||
| GenBank mRNA | JU496883.1 | - | 123 | 104 | ||
| JV727624.1 | - | 105 | 104 | |||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015124738.2 transcripts.
Set Membership: |
|

ZNF432
Genomic Map