Assay Details
| Gene Symbol: ZNF432 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015124738.2 | XP_014980224.1 | 653 | 76 | ||
| XM_015124740.2 | XP_014980226.1 | 492 | 76 | |||
| GenBank mRNA | JU496883.1 | - | 556 | 76 | ||
| JV727624.1 | - | 538 | 76 | |||
More Information
Set Membership: |
|

ZNF432
Genomic Map