Assay Details
| Gene Symbol: ANGPTL8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015122931.2 | XP_014978417.1 | 479 | 62 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

ANGPTL8| Gene Symbol: ANGPTL8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015122931.2 | XP_014978417.1 | 479 | 62 | ||
Set Membership: |
|