Assay Details
| Gene Symbol: THAP10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001089200.4 | XP_001089200.1 | 665 | 65 | ||
| GenBank mRNA | JU335529.1 | - | 463 | 65 | ||
| JV704142.1 | - | 460 | 65 | |||
More Information
Set Membership: |
|

THAP10
Genomic Map