Assay Details
| Gene Symbol: THAP10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001089200.4 | XP_001089200.1 | 818 | 95 | ||
| GenBank mRNA | JU335529.1 | - | 616 | 95 | ||
| JV704142.1 | - | 613 | 95 | |||
More Information
Set Membership: |
|

THAP10
Genomic Map