Assay Details
Gene Symbol: TOMM34 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015148947.2 | XP_015004433.1 | 884 | 75 | ||
XM_015148948.2 | XP_015004434.1 | 899 | 75 | |||
GenBank mRNA | JU332654.1 | - | 786 | 75 | ||
JU472427.1 | - | 775 | 75 | |||
JV046020.1 | - | 804 | 75 | |||
JV565058.1 | - | 786 | 75 | |||
JV703782.1 | - | 799 | 75 |

More Information
Set Membership: |
![]() ![]() ![]() |
