Assay Details
| Gene Symbol: PGAM5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015109735.2 | XP_014965221.2 | 784 | 54 | ||
| XM_028830238.1 | XP_028686071.1 | 781 | 54 | |||
| GenBank mRNA | JU327809.1 | - | 612 | 54 | ||
| JU474583.1 | - | 776 | 54 | |||
| JU474584.1 | - | 773 | 54 | |||
| JV053879.1 | - | 696 | 54 | |||
| JV636005.1 | - | 743 | 54 | |||
More Information
Set Membership: |
|

PGAM5
Genomic Map