Assay Details
| Gene Symbol: OSGIN1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015126698.2 | XP_014982184.2 | 665 | 56 | ||
| XM_015126699.2 | XP_014982185.2 | 606 | 56 | |||
| XM_015126700.2 | XP_014982186.2 | 615 | 56 | |||
| GenBank mRNA | JV579827.1 | - | 638 | 56 | ||
More Information
Set Membership: |
|

OSGIN1
Genomic Map