Assay Details
Gene Symbol: SEPTIN9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015120460.2 | XP_014975946.2 | 2023 | 60 | ||
XM_015120462.2 | XP_014975948.2 | 1747 | 60 | |||
XM_015120463.2 | XP_014975949.2 | 2185 | 60 | |||
XM_015120464.2 | XP_014975950.2 | 2407 | 60 | |||
XM_015120465.2 | XP_014975951.1 | 1162 | 60 | |||
XM_015120466.2 | XP_014975952.1 | 1457 | 60 | |||
XM_015120467.2 | XP_014975953.1 | 1266 | 60 | |||
XM_015120468.2 | XP_014975954.1 | 1059 | 60 | |||
XM_028836849.1 | XP_028692682.1 | 1492 | 60 | |||
XM_028836850.1 | XP_028692683.1 | 997 | 60 | |||
GenBank mRNA | JU322456.1 | - | 1750 | 60 | ||
JV045986.1 | - | 1802 | 60 | |||
JV045987.1 | - | 1807 | 60 | |||
JV527780.1 | - | 1651 | 60 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
