Assay Details
| Gene Symbol: SEPTIN9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015120460.2 | XP_014975946.2 | 1116 | 113 | ||
| XM_015120462.2 | XP_014975948.2 | 840 | 113 | |||
| XM_015120463.2 | XP_014975949.2 | 1278 | 113 | |||
| XM_015120464.2 | XP_014975950.2 | 1500 | 113 | |||
| GenBank mRNA | JU322456.1 | - | 843 | 113 | ||
| JV045986.1 | - | 900 | 113 | |||
| JV045987.1 | - | 900 | 113 | |||
| JV527780.1 | - | 744 | 113 | |||
More Information
Set Membership: |
|

SEPTIN9
Genomic Map