Assay Details
| Gene Symbol: ISCA2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001261007.1 | NP_001247936.1 | 309 | 110 | ||
| GenBank mRNA | JU322820.1 | - | 317 | 110 | ||
| JU474273.1 | - | 309 | 110 | |||
| JV605565.1 | - | 322 | 110 | |||
More Information
Set Membership: |
|

ISCA2
Genomic Map