Assay Details
| Gene Symbol: HTR2B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015111331.2 | XP_014966817.1 | 1231 | 98 | ||
| XM_028831205.1 | XP_028687038.1 | 1476 | 98 | |||
| GenBank mRNA | JV059701.1 | - | 203 | 98 | ||
More Information
Set Membership: |
|

HTR2B
Genomic Map