Assay Details
| Gene Symbol: ZNF771 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001100716.4 | XP_001100716.2 | 530 | 143 | ||
| XM_028840928.1 | XP_028696761.1 | 672 | 143 | |||
| XM_028840929.1 | XP_028696762.1 | 264 | 143 | |||
| GenBank mRNA | JU334866.1 | - | 195 | 143 | ||
| JV601196.1 | - | 251 | 143 | |||
| JV716487.1 | - | 229 | 143 | |||
More Information
Set Membership: |
|

ZNF771
Genomic Map