Assay Details
| Gene Symbol: SHISA8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015150476.2 | XP_015005962.2 | 958 | 83 | ||
| XM_028827652.1 | XP_028683485.1 | 558 | 83 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

SHISA8| Gene Symbol: SHISA8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015150476.2 | XP_015005962.2 | 958 | 83 | ||
| XM_028827652.1 | XP_028683485.1 | 558 | 83 | |||