Assay Details
| Gene Symbol: SHISA8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015150476.2 | XP_015005962.2 | 1523 | 76 | ||
| XM_028827652.1 | XP_028683485.1 | 1003 | 76 | |||
More Information
Set Membership: |
|

SHISA8
Genomic Map