Assay Details
| Gene Symbol: ZNF613 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015124717.2 | XP_014980203.2 | 805 | 105 | ||
| XM_015124718.2 | XP_014980204.2 | 2394 | 105 | |||
| XM_015124719.2 | XP_014980205.2 | 802 | 105 | |||
| XM_015124721.2 | XP_014980207.2 | 723 | 105 | |||
| XM_015124722.2 | XP_014980208.2 | 387 | 105 | |||
| XM_028839359.1 | XP_028695192.1 | 513 | 105 | |||
| XM_028839360.1 | XP_028695193.1 | 597 | 105 | |||
| XM_028839361.1 | XP_028695194.1 | 305 | 105 | |||
| XM_028839362.1 | XP_028695195.1 | 594 | 105 | |||
| GenBank mRNA | JU348689.1 | - | 188 | 105 | ||
| JU493718.1 | - | 429 | 105 | |||
More Information
Set Membership: |
|

ZNF613
Genomic Map