Assay Details
| Gene Symbol: OSM | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001194474.3 | NP_001181403.1 | 222 | 96 | ||
| XM_015150017.2 | XP_015005503.1 | 3795 | 96 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

OSM| Gene Symbol: OSM | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001194474.3 | NP_001181403.1 | 222 | 96 | ||
| XM_015150017.2 | XP_015005503.1 | 3795 | 96 | |||
Set Membership: |
|