Assay Details
| Gene Symbol: TNFAIP8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001261083.1 | NP_001248012.1 | 148 | 83 | ||
| GenBank mRNA | JU473569.1 | - | 203 | 83 | ||
| JV044110.1 | - | 148 | 83 | |||
| JV635588.1 | - | 156 | 83 | |||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001261083.1 transcripts.
Set Membership: |
|

TNFAIP8
Genomic Map