Assay Details
| Gene Symbol: ACKR3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001261729.1 | NP_001248658.1 | 1517 | 101 | ||
| XM_015111240.2 | XP_014966726.1 | 1428 | 101 | |||
| XM_015111241.2 | XP_014966727.1 | 1631 | 101 | |||
| XM_028830379.1 | XP_028686212.1 | 1877 | 101 | |||
| GenBank mRNA | JU332120.1 | - | 1407 | 101 | ||
| JV043892.1 | - | 1517 | 101 | |||
More Information
Set Membership: |
|

ACKR3
Genomic Map