Assay Details
| Gene Symbol: FJX1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015114526.2 | XP_014970012.2 | 2013 | 53 | ||
| GenBank mRNA | JU325850.1 | - | 120 | 53 | ||
| JV678927.1 | - | 1010 | 53 | |||
More Information
Set Membership: |
|

FJX1
Genomic Map