Assay Details
| Gene Symbol: CDR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015128332.2 | XP_014983818.1 | 569 | 91 | ||
| GenBank mRNA | JU330112.1 | - | 125 | 91 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

CDR1| Gene Symbol: CDR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015128332.2 | XP_014983818.1 | 569 | 91 | ||
| GenBank mRNA | JU330112.1 | - | 125 | 91 | ||
Set Membership: |
|