Assay Details
| Gene Symbol: SPESP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001084797.4 | XP_001084797.1 | 159 | 61 | ||
| XM_015142556.2 | XP_014998042.1 | 159 | 61 | |||
| GenBank mRNA | JV711127.1 | - | 120 | 61 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_001084797.4, XM_015142556.2 transcripts.
Set Membership: |
|

SPESP1
Genomic Map