Assay Details
| Gene Symbol: HSPA2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_028851610.1 | XP_028707443.1 | 2003 | 82 | ||
| GenBank mRNA | AY369825.1 | - | 515 | 82 | ||
| JU320353.1 | - | 1976 | 82 | |||
| JV682390.1 | - | 1772 | 82 | |||
More Information
Set Membership: |
|

HSPA2
Genomic Map