Assay Details
| Gene Symbol: TMEM41B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002799640.3 | XP_002799686.2 | 796 | 94 | ||
| GenBank mRNA | JU322882.1 | - | 643 | 94 | ||
| JU472956.1 | - | 633 | 94 | |||
| JV045785.1 | - | 570 | 94 | |||
More Information
Set Membership: |
|

TMEM41B
Genomic Map