Assay Details
| Gene Symbol: FAM32A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015123210.2 | XP_014978696.1 | 295 | 68 | ||
| GenBank mRNA | JU333903.1 | - | 235 | 68 | ||
| JU475090.1 | - | 235 | 68 | |||
| JU475091.1 | - | 235 | 68 | |||
| JV046702.1 | - | 236 | 68 | |||
More Information
Set Membership: |
|

FAM32A
Genomic Map