Assay Details
| Gene Symbol: DLEU7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001104961.3 | XP_001104961.1 | 1297 | 109 | ||
| GenBank mRNA | JU340333.1 | - | 219 | 109 | ||
| JV709485.1 | - | 225 | 109 | |||
More Information
Set Membership: |
|

DLEU7
Genomic Map