Assay Details
| Gene Symbol: MMRN2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015147080.2 | XP_015002566.2 | 734 | 62 | ||
| XM_015147081.2 | XP_015002567.2 | 728 | 62 | |||
| GenBank mRNA | JV520685.1 | - | 338 | 62 | ||
| JV723603.1 | - | 209 | 62 | |||
More Information
Set Membership: |
|

MMRN2
Genomic Map