Assay Details
| Gene Symbol: TMC8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_028836852.1 | XP_028692685.1 | 2121 | 67 | ||
| XM_028836853.1 | XP_028692686.1 | 2121 | 67 | |||
| XM_028836854.1 | XP_028692687.1 | 1764 | 67 | |||
| GenBank mRNA | JU474404.1 | - | 1725 | 67 | ||
| JU480856.1 | - | 1102 | 67 | |||
More Information
Set Membership: |
|

TMC8
Genomic Map