Assay Details
| Gene Symbol: TTC9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015144030.2 | XP_014999516.1 | 1124 | 84 | ||
| GenBank mRNA | JU326310.1 | - | 415 | 84 | ||
| JV678561.1 | - | 532 | 84 | |||
More Information
Set Membership: |
|

TTC9
Genomic Map