Assay Details
| Gene Symbol: NYNRIN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015143465.2 | XP_014998951.1 | 506 | 55 | ||
| XM_028851461.1 | XP_028707294.1 | 375 | 55 | |||
| GenBank mRNA | JV043216.1 | - | 258 | 55 | ||
| JV635397.1 | - | 380 | 55 | |||
More Information
Set Membership: |
|

NYNRIN
Genomic Map