Assay Details
| Gene Symbol: SEPTIN9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015120463.2 | XP_014975949.2 | 638 | 65 | ||
| GenBank mRNA | JU322456.1 | - | 203 | 65 | ||
| JV045986.1 | - | 260 | 65 | |||
| JV045987.1 | - | 260 | 65 | |||
More Information
Set Membership: |
|

SEPTIN9
Genomic Map