Assay Details
Gene Symbol: SEPTIN9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015120460.2 | XP_014975946.2 | 1872 | 57 | ||
XM_015120462.2 | XP_014975948.2 | 1596 | 57 | |||
XM_015120463.2 | XP_014975949.2 | 2034 | 57 | |||
XM_015120464.2 | XP_014975950.2 | 2256 | 57 | |||
XM_015120465.2 | XP_014975951.1 | 1011 | 57 | |||
XM_015120466.2 | XP_014975952.1 | 1306 | 57 | |||
XM_015120467.2 | XP_014975953.1 | 1115 | 57 | |||
XM_015120468.2 | XP_014975954.1 | 908 | 57 | |||
XM_028836849.1 | XP_028692682.1 | 1341 | 57 | |||
XM_028836850.1 | XP_028692683.1 | 846 | 57 | |||
GenBank mRNA | JU322456.1 | - | 1599 | 57 | ||
JV045986.1 | - | 1651 | 57 | |||
JV045987.1 | - | 1656 | 57 | |||
JV527780.1 | - | 1500 | 57 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
