Assay Details
| Gene Symbol: OLFML1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015114956.2 | XP_014970442.1 | 682 | 67 | ||
| GenBank mRNA | JU326744.1 | - | 73 | 67 | ||
| JV045058.1 | - | 529 | 67 | |||
| JV639223.1 | - | 567 | 67 | |||
More Information
Set Membership: |
|

OLFML1
Genomic Map