Assay Details
| Gene Symbol: ZNF771 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001100716.4 | XP_001100716.2 | 383 | 59 | ||
| GenBank mRNA | JU334866.1 | - | 48 | 59 | ||
| JV601196.1 | - | 104 | 59 | |||
| JV716487.1 | - | 82 | 59 | |||
More Information
Set Membership: |
|

ZNF771
Genomic Map