Assay Details
| Gene Symbol: MRPL54 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015122474.2 | XP_014977960.1 | 179 | 68 | ||
| GenBank mRNA | JU332832.1 | - | 146 | 68 | ||
| JV568016.1 | - | 150 | 68 | |||
| JV667243.1 | - | 131 | 68 | |||
More Information
Set Membership: |
|

MRPL54
Genomic Map