Assay Details
| Gene Symbol: ZFP30 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015123626.2 | XP_014979112.2 | 3441 | 117 | ||
| XM_015123628.2 | XP_014979114.2 | 3031 | 117 | |||
| XM_015123631.2 | XP_014979117.2 | 3438 | 117 | |||
| XM_015123632.2 | XP_014979118.2 | 3349 | 117 | |||
| XM_028839054.1 | XP_028694887.1 | 3121 | 117 | |||
| GenBank mRNA | JU327856.1 | - | 1614 | 117 | ||
| JV709746.1 | - | 2034 | 117 | |||
| JV709747.1 | - | 2643 | 117 | |||
More Information
Set Membership: |
|

ZFP30
Genomic Map