Assay Details
| Gene Symbol: ZNF444 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015125120.2 | XP_014980606.2 | 1897 | 66 | ||
| XM_015125122.2 | XP_014980608.2 | 1982 | 66 | |||
| XM_015125123.2 | XP_014980609.2 | 1830 | 66 | |||
| XM_015125124.2 | XP_014980610.2 | 1953 | 66 | |||
| XM_015125125.2 | XP_014980611.2 | 1870 | 66 | |||
| XM_015125127.2 | XP_014980613.2 | 2104 | 66 | |||
| XM_015125128.2 | XP_014980614.2 | 1894 | 66 | |||
| XM_028840169.1 | XP_028696002.1 | 1875 | 66 | |||
| XM_028840170.1 | XP_028696003.1 | 2251 | 66 | |||
| GenBank mRNA | JV586395.1 | - | 424 | 66 | ||
More Information
Set Membership: |
|

ZNF444
Genomic Map