Assay Details
| Gene Symbol: NAGLU | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015119713.2 | XP_014975199.2 | 1900 | 81 | ||
| GenBank mRNA | JV061835.1 | - | 988 | 81 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

NAGLU| Gene Symbol: NAGLU | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015119713.2 | XP_014975199.2 | 1900 | 81 | ||
| GenBank mRNA | JV061835.1 | - | 988 | 81 | ||
Set Membership: |
|