Assay Details
| Gene Symbol: FIGNL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015133495.2 | XP_014988981.2 | 214 | 103 | ||
| XM_015133497.2 | XP_014988983.2 | 860 | 103 | |||
| GenBank mRNA | JV068351.1 | - | 118 | 103 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015133495.2, XM_015133497.2 transcripts.
Set Membership: |
|

FIGNL1
Genomic Map