Assay Details
| Gene Symbol: SPRY4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001095682.3 | XP_001095682.1 | 4452 | 80 | ||
| XM_028849810.1 | XP_028705643.1 | 5168 | 80 | |||
| GenBank mRNA | JU349104.1 | - | 2721 | 80 | ||
| JV724409.1 | - | 2923 | 80 | |||
More Information
Set Membership: |
|

SPRY4
Genomic Map