Assay Details
| Gene Symbol: TUBGCP5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_028850598.1 | XP_028706431.1 | 2969 | 88 | ||
| GenBank mRNA | JU332801.1 | - | 2846 | 88 | ||
| JU472779.1 | - | 2843 | 88 | |||
| JV638134.1 | - | 2857 | 88 | |||
| Gene Symbol: LOC114675405 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_028842664.1 | XP_028698497.1 | 2105 | 88 | ||
| XM_028842665.1 | XP_028698498.1 | 2018 | 88 | |||
| XM_028842666.1 | XP_028698499.1 | 2105 | 88 | |||
| XM_028842667.1 | XP_028698500.1 | 1922 | 88 | |||
| XM_028842668.1 | XP_028698501.1 | 2235 | 88 | |||
| XM_028842669.1 | XP_028698502.1 | 1902 | 88 | |||
| XM_028842670.1 | XP_028698503.1 | 1733 | 88 | |||
| XM_028842671.1 | XP_028698504.1 | 1733 | 88 | |||
| GenBank mRNA | JV570350.1 | - | 1996 | 88 | ||
More Information
Set Membership: |
|

TUBGCP5
Genomic Map