Assay Details
| Gene Symbol: PATZ1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002798338.3 | XP_002798384.3 | 2068 | 64 | ||
| XM_028827826.1 | XP_028683659.1 | 1985 | 64 | |||
| XM_028827827.1 | XP_028683660.1 | 1985 | 64 | |||
| GenBank mRNA | JU333743.1 | - | 1379 | 64 | ||
| JU333744.1 | - | 1379 | 64 | |||
| JU333745.1 | - | 1379 | 64 | |||
| JU472373.1 | - | 1806 | 64 | |||
| JU472375.1 | - | 1806 | 64 | |||
| JV044273.1 | - | 1703 | 64 | |||
| JV044275.1 | - | 1703 | 64 | |||
| JV637479.1 | - | 1954 | 64 | |||
More Information
Set Membership: |
|

PATZ1
Genomic Map