Assay Details
| Gene Symbol: CASP6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015139016.2 | XP_014994502.2 | 1718 | 75 | ||
| XM_015139017.2 | XP_014994503.2 | 728 | 75 | |||
| XM_015139018.2 | XP_014994504.2 | 1698 | 75 | |||
| XM_015139019.2 | XP_014994505.2 | 708 | 75 | |||
| GenBank mRNA | JU349307.1 | - | 576 | 75 | ||
| JU477441.1 | - | 698 | 75 | |||
| JV636402.1 | - | 713 | 75 | |||
More Information
Set Membership: |
|

CASP6
Genomic Map