Assay Details
| Gene Symbol: CCZ1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015133273.2 | XP_014988759.1 | 1664 | 91 | ||
| XM_028845138.1 | XP_028700971.1 | 1099 | 91 | |||
| XM_028845139.1 | XP_028700972.1 | 1071 | 91 | |||
| GenBank mRNA | JU334942.1 | - | 1123 | 91 | ||
| JU475946.1 | - | 1132 | 91 | |||
| JU475947.1 | - | 1176 | 91 | |||
| JV584224.1 | - | 1094 | 91 | |||
| JV584225.1 | - | 1080 | 91 | |||
More Information
Set Membership: |
|

CCZ1
Genomic Map